Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ L-Quisqualic acid
GREENER_CHOICE

Catalog No. 018810
Change view
Click to view available options
Quantity:
1 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-881-0 10 mg
01-881 1 mg
01-885-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 01-881-0 Supplier Tocris Bioscience™ Supplier No. 0188/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

AMPA agonist; also group I mGlu agonist

L-Quisqualic acid is a glutamate receptor agonist acting at AMPA receptors and metabotropic glutamate receptors positively linked to phosphoinositide hydrolysis. L-Quisqualic acid sensitizes neurons in hippocampus to depolarization by L-AP6 (the so called 'quis' effect).

Chemical Identifiers

CAS 52809-07-1
Molecular Formula C5H7N3O5

Specifications

Quantity 10 mg
Formula Weight 189.13
Purity >99%
Molecular Weight (g/mol) 189.13
Chemical Name or Material L-Quisqualic acid
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.