Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
L-Selectin/CD62L Antibody (018), Alexa Fluor™ 350

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289996AF350
Description
L-Selectin/CD62L Monoclonal specifically detects L-Selectin/CD62L in Human samples. It is validated for ELISA, Sandwich ELISA Detection.Specifications
L-Selectin/CD62L | |
Monoclonal | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
18 | |
ELISA, Sandwich ELISA Detection | |
CD62L, CD62L antigen, gp90-MEL, hLHRc, LAM-1, LAM1LECAM1, Leu-8, LEU8, Leukocyte adhesion molecule 1, Leukocyte surface antigen Leu-8, Leukocyte-endothelial cell adhesion molecule 1, LNHRTQ1, LSEL, L-selectin, Lyam-1, LYAM1CD62 antigen-like family member L, Lymph node homing receptor, lymphocyte adhesion molecule 1, pln homing receptor, PLNHR, selectin L | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human L-Selectin/CD62L (P14151-1; Met 1-Asn 332). | |
0.1 mL | |
Adaptive Immunity, B Cell Development and Differentiation Markers, Cardiovascular Biology, Glycobiology, Immunology, Lipid and Metabolism, Myeloid derived Suppressor Cell, Signal Transduction, Stem Cells | |
6402 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction