Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
L-Selectin/CD62L Antibody (FMC46), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10065388C
Description
L-Selectin/CD62L Monoclonal specifically detects L-Selectin/CD62L in Human, Bovine, Canine, Primate samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
L-Selectin/CD62L | |
Monoclonal | |
DyLight 650 | |
50 mM sodium borate with 0.05% sodium azide | |
SELL | |
PHA stimulated lymphoblasts. | |
0.1 mL | |
Primary | |
NB100-65388 recognizes human CD62L, also known a L-selectin, a 74-95kDa member of the selectin family of adhesion receptors, which acts as a ligand for both CD62-P (P-selectin) and CD62E (E-selectin). Human CD62L is constitutively expressed on most leucocytes including monocytes, granulocytes, lymphocytes, NK cells, bone marrow myeloid progenitor cells and on a subset of thymocytes. CD62L plays an important role in leucocyte tethering and rolling on the endothelial cell surface and for the homing of naive lymphocytes to lymph nodes and Peyers patches via HEV. Neutrophils require a constant supply of this molecule on the cell surface for migration into peripheral tissues and adhesion to activated endothelium at sites of inflammation, where CD62L is rapidly shed as soluble L-selectin, but surface expression still remains. The expression of CD62L is down regulated on lymphocytes and neutrophils by PMA stimulation. | |
Store at 4°C in the dark. |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
FMC46 | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen | |
CD62L, CD62L antigen, gp90-MEL, hLHRc, LAM-1, LAM1LECAM1, Leu-8, LEU8, Leukocyte adhesion molecule 1, Leukocyte surface antigen Leu-8, Leukocyte-endothelial cell adhesion molecule 1, LNHRTQ1, LSEL, L-selectin, Lyam-1, LYAM1CD62 antigen-like family member L, Lymph node homing receptor, lymphocyte adhesion molecule 1, pln homing receptor, PLNHR, selectin L | |
Mouse | |
Protein G purified | |
Adaptive Immunity, B Cell Development and Differentiation Markers, Cardiovascular Biology, Glycobiology, Immunology, Lipid and Metabolism, Myeloid derived Suppressor Cell, Signal Transduction, Stem Cells | |
6402.0 | |
Human, Bovine, Canine, Primate | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction