Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Selenocystine, 95%, Thermo Scientific Chemicals

Catalog No. p-191786
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 29621-88-3 | C6H12N2O4Se2 | 334.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29621-88-3
Molecular Formula C6H12N2O4Se2
Molecular Weight (g/mol) 334.11
MDL Number MFCD00800971
InChI Key JULROCUWKLNBSN-IMJSIDKUSA-N
Synonym l-selenocystine, seleno-l-cystine, l-3,3'-diselenodialanine, alanine, 3,3'-diselenodi-, l, selenocystine, l, l-alanine,3,3'-diselenobis, 2r-2-amino-3-2r-2-amino-2-carboxyethyl diselanyl propanoic acid, l-selenocystine monohydrate, selenocystine, l-isomer
PubChem CID 207306
IUPAC Name (2R)-2-amino-3-[[(2R)-2-amino-2-carboxyethyl]diselanyl]propanoic acid
SMILES N[C@@H](C[Se][Se]C[C@H](N)C(O)=O)C(O)=O

Specifications

Melting Point 212°C to 215°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 250 mg
Solubility Information Solubility in water: soluble in water
Formula Weight 334.09
Percent Purity 98%
Physical Form Powder
Chemical Name or Material L-Selenocystine, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Toxic if inhaled.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RTECSNumber AY6032000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.