Alfa Aesar™ L-Serine, 99%

Manufacturer: Alfa Aesar™ A1117914

Quantity 25g

 View more versions of this product

Catalog No. AAA1117914

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 56-45-1
    Molecular Formula C3H7NO3
    Formula Weight 105.09
    MDL Number MFCD00064224
    Beilstein 1721404
    Merck Index 14,8460
    Synonym (S)-2-Amino-3-hydroxypropionic acid; H-Ser-OH
    Chemical Name or Material L-Serine

    Specifications

    Formula Weight 105.09
    TSCA TSCA
    Molecular Formula C3H7NO3
    CAS 56-45-1
    Merck Index 14,8460
    Melting Point 214° to 224°C (dec.)
    Optical Rotation +15° (c=9 in 1N HCl)
    MDL Number MFCD00064224
    Beilstein 1721404
    Synonym (S)-2-Amino-3-hydroxypropionic acid; H-Ser-OH
    Quantity 25g
    RTECS Number VT8100000
    Chemical Name or Material L-Serine
    Assay 99%

    Description

    Applications
    L-serine is used as a dietary supplement to improve health under certain conditions of malnutrition or physical stress, as a cosmetic incredient, as ingredients for cell culture media. It is used as a precursor to several amino acids, including glycine, cysteine and other metabolites such as sphingolipids and folate. It is used in bio-synthesis of purines and pyrimidines. It plays a vital role in catalytic function of enzymes. Further, L-serine and its metabolic products are essential for cell proliferation and specific functions of the central nervous system. .

    Solubility
    Soluble in water. Insoluble in ethanol, ether and benzene.

    Notes
    Store in a cool place. Incompatible with strong oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit