Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Serine, FCC, 98.5-101.5%, Spectrum™ Chemical
SDP

Catalog No. 18603980 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Poly Bottle
Poly Pail

Catalog No. 18-603-980

Supplier: Spectrum Chemical Manufacturing Corporation S1021100GM

Only null left
Add to Cart
Add to Cart

S1021, 56-45-1, C3H7NO3

Spectrum™ Chemical L-Serine, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 56-45-1
Molecular Formula C3H7NO3
Molecular Weight (g/mol) 105.09
InChI Key MTCFGRXMJLQNBG-UHFFFAOYNA-N
IUPAC Name 2-amino-3-hydroxypropanoic acid
SMILES NC(CO)C(O)=O

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 0.3%
Packaging Poly Bottle
Quantity 100 g
Optical Rotation +13.6° to +160°
Percent Purity 98.5 to 101.5%
Grade FCC
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.