Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L(+)-Tartaric acid, 99+%, Acros Organics

Click to view available options
Quantity:
500g
2.5kg
10kg
Packaging:
Plastic bottle
Plastic drum
87-69-4
C4H6O6
150.086
FEWJPZIEWOKRBE-JCYAYHJZSA-N
l-tartaric acid,l-+-tartaric acid,l +-tartaric acid,2r,3r-2,3-dihydroxysuccinic acid,tartaric acid,2r,3r-2,3-dihydroxybutanedioic acid,r,r-tartaric acid,+-l-tartaric acid,dextrotartaric acid,l-threaric acid
444305
CHEBI:15671
(2R,3R)-2,3-dihydroxybutanedioic acid
C(C(C(=O)O)O)(C(=O)O)O
87-69-4
C4H6O6
150.086
FEWJPZIEWOKRBE-JCYAYHJZSA-N
l-tartaric acid,l-+-tartaric acid,l +-tartaric acid,2r,3r-2,3-dihydroxysuccinic acid,tartaric acid,2r,3r-2,3-dihydroxybutanedioic acid,r,r-tartaric acid,+-l-tartaric acid,dextrotartaric acid,l-threaric acid
444305
CHEBI:15671
(2R,3R)-2,3-dihydroxybutanedioic acid
C(C(C(=O)O)O)(C(=O)O)O
Assay Percent Range 99+%
Calcium (Ca) 200ppm max.
Linear Formula HO2CCH(OH)CH(OH)CO2H
Chlorine (Cl) 100ppm max.
Formula Weight 150.09
Physical Form Crystalline Powder or Crystals
Percent Purity ≥99%
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Loss on Drying 0.5% (1g, 105°C) max.
Packaging Plastic drum
Solubility Solubility in water: 1390g/L (20°C). Other solubilities: soluble in methanol, ethanol, propanol and, glycerol, 4g/L ether, insoluble in chloroform
Specific Rotation + 12.00
Sulfate 150ppm max.
Sulfated Ash 0.1% max.
Specific Rotation Condition + 12.00 (20.00°C c=20,H2O)
Melting Point 168.0°C to 172.0°C
Color White or Colorless
Quantity 10kg
Flash Point 210°C
Chemical Name or Material L(+)-Tartaric acid
Show More Show Less