Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-(+)-Tartaric acid, ACS

Catalog No. aa3640522
Click to view available options
Quantity:
100 g
500 g
2 kg

Useful as a resolving agent | CAS: 87-69-4 | C4H6O6 | 150.09 g/mol

L-(+)-Tartaric acid is widely utilized in pharmaceutical industries. It is used in soft drinks, confectionaries, food products, gelatin desserts and buffering agents. It forms a compound, TiCl2(O-i-Pr)2 with Diels-Alder catalyst and act as a chelate agent in metal industries. Owing to its efficient chelating property towards metal ions, it is used in the farming and metal industries for complexing micronutrients and for cleaning metal surfaces, respectively.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-69-4
Molecular Formula C4H6O6
Molecular Weight (g/mol) 150.09
MDL Number MFCD00064207
InChI Key FEWJPZIEWOKRBE-UHFFFAOYNA-N
Synonym l-tartaric acid, l-+-tartaric acid, l +-tartaric acid, 2r,3r-2,3-dihydroxysuccinic acid, tartaric acid, 2r,3r-2,3-dihydroxybutanedioic acid, r,r-tartaric acid, +-l-tartaric acid, dextrotartaric acid, l-threaric acid
PubChem CID 444305
ChEBI CHEBI:15671
SMILES OC(C(O)C(O)=O)C(O)=O

Specifications

Melting Point 168°C to 170°C
Beilstein 1725147
Merck Index 14,9070
Solubility Information Soluble in water,methanol and acetone.
IUPAC Name 2,3-dihydroxybutanedioic acid
Formula Weight 150.09
Grade ACS
Density 1.76
Quantity 100 g
Physical Form Granular
Chemical Name or Material L-(+)-Tartaric Acid
Show More Show Less
Hazard Category H318-H500
Hazard Statement GHS H Statement
H318
Precautionary Statement P280-P305+P351+P338-P310
EINECSNumber 201-766-0
RTECSNumber WW7875000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.