Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-(+)-Tartaric Acid (Fine Granular/NF/FCC), Fisher Chemical

Catalog No. a313500 Shop All Fisherbrand Products
Click to view available options
:
Poly Bottle; 500g

Specifications

pH 1.6
Melting Point 168°C
CAS 87-69-4
Color White
Molecular Formula C4H6O6
Linear Formula HOOC(CHOH)2COOH
MDL Number MFCD00064207
Synonym l-tartaric acid, l-+-tartaric acid, l +-tartaric acid, 2r,3r-2,3-dihydroxysuccinic acid, tartaric acid, 2r,3r-2,3-dihydroxybutanedioic acid, r,r-tartaric acid, +-l-tartaric acid, dextrotartaric acid, l-threaric acid
InChI Key FEWJPZIEWOKRBE-UHFFFAOYNA-N
SMILES OC(C(O)C(O)=O)C(O)=O
IUPAC Name 2,3-dihydroxybutanedioic acid
Molecular Weight (g/mol) 150.09
PubChem CID 444305
ChEBI CHEBI:15671
Formula Weight 150.09
Percent Purity 99.7 to 100.5% (NF)
Grade FCC/NF
Quantity 500 g
Ignition Residue 0.05% max. (FCC)
Loss on Drying 0.5% max. (FCC), 0.5% max. (FCC/NF), 0.5% max. (NF)
Packaging Poly Bottle
Physical Form Solid
Residue after Ignition 0.1% max. (NF)
Vapor Pressure <0.1 mbar at 20°C
Chemical Name or Material L-(+)-Tartaric Acid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.