Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Thyroxine, 98%

Catalog No. p-7042461
Click to view available options
Quantity:
1 g
5 g

CAS: 51-48-9 | C15H11I4NO4 | 776.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51-48-9
Molecular Formula C15H11I4NO4
Molecular Weight (g/mol) 776.87
MDL Number MFCD00002595
InChI Key XUIIKFGFIJCVMT-UHFFFAOYNA-N
Synonym l-thyroxine, levothyroxine, thyroxine, thyroxin, synthroid, tetraiodothyronine, thyrax, levothyroxin, thyratabs, thyreoideum
PubChem CID 5819
ChEBI CHEBI:18332
IUPAC Name 2-amino-3-[4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]propanoic acid
SMILES NC(CC1=CC(I)=C(OC2=CC(I)=C(O)C(I)=C2)C(I)=C1)C(O)=O

Specifications

Melting Point 235°C to 236°C
Color Brown to White
Odor Odorless
Quantity 1 g
Beilstein 2228515
Sensitivity Light sensitive
Merck Index 14,9415
Solubility Information Dissolves in 4M ammonium hydroxide in Methanol at 50mg/ml
Formula Weight 776.87
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material L-Thyroxine
Show More Show Less
Hazard Category H302
Precautionary Statement P264-P270-P301+P312-P330-P501c
EINECSNumber 200-101-1
RTECSNumber YP2833500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.