Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L1/ORF2 Polyclonal Antibody, Rockland™
SDP

Catalog No. 501991312
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-1312 100 μg
50-199-1313 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-1312 Supplier Rockland Immunochemicals Supplier No. 200901P11
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Anti-L1/ORF2 Antibody is directed against three regions within the endonuclease domain of L1 ORF2 protein.Reactivity occurs against mouse L1/ORF2 protein and is useful in determining its presence in various assays. A BLAST analysis shows reactivity with L1/ORF2 proteins from mouse sources based on 100% homology with the immunizing sequences. Cross reactivity against L1/ORF2 from other sources is unknown.

The L1 (LINE-1), or Long INterspersed Element , retrotransposon ORF2 is the most common open reading frame in the human genome, present in various forms in many thousands of copies. This large family of proteins includes magnesium dependent endonucleases and a large number of phosphatases involved in intracellular signaling. Both intact ORF1 and ORF2 are absolutely required for autonomous retro-transposition. ORF2 encodes an endonuclease, reverse transcriptase, and zinc knuckle domains. The expression of ORF2 appears to be tightly regulated except in germ line tissues, embryonic tissues, and certain cancers including teratomas, testicular cancers, and leukemias. This antibody is intended for use in studying control of L1 retrotransposons.

Specifications

Antigen L1/ORF2
Applications ELISA
Classification Polyclonal
Concentration 1.21 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Host Species Chicken
Immunogen This IgY fraction antibody was prepared from eggs of chickens laid after repeated immunizations with three synthetic peptides conjugated to keyhole limpet hemocyanin (KLH). The peptides correspond to regions within the endonuclease domain of L1/ORF2 protein.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.