Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
L1CAM Antibody (2702C), Alexa Fluor™ 532, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB7773X
Description
L1CAM Monoclonal antibody specifically detects L1CAM in Human samples. It is validated for Western Blot, Western Blot, Flow Cytometry, ImmunocytochemistrySpecifications
L1CAM | |
Monoclonal | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, Immunocytochemistry | |
2702C | |
Western Blot, Simple Western, Flow Cytometry, Immunocytochemistry | |
antigen identified by monoclonal R1, CAML1N-CAML1, CD171, CD171 antigen, HSAS, HSAS1, L1 cell adhesion molecule, MASA, MIC5, N-CAM-L1, NCAM-L1, neural cell adhesion molecule L1, S10, SPG1 | |
Mouse myeloma cell line NS0 derived Human L1CAM, (Ile20Glu1120) and (Arg864Glu1120), Accession # CAA42508 | |
0.1 mL | |
Cellular Markers, Glia Markers, Neuroscience, Phospho Specific | |
3897 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction