Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
L1CAM Antibody (84307), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB7772PEATT594
Description
L1CAM Monoclonal antibody specifically detects L1CAM in Human, Mouse samples. It is validated for Flow CytometrySpecifications
L1CAM | |
Monoclonal | |
PE/ATTO 594 | |
PBS | |
Mouse | |
Protein A or G purified from ascites | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Flow Cytometry | |
84307 | |
Flow Cytometry | |
antigen identified by monoclonal R1, CAML1N-CAML1, CD171, CD171 antigen, HSAS, HSAS1, L1 cell adhesion molecule, MASA, MIC5, N-CAM-L1, NCAM-L1, neural cell adhesion molecule L1, S10, SPG1 | |
Mouse myeloma cell line NS0-derived recombinant human/mouse L1CAM, Met1-Glu1120, Accession # CAA42508 | |
0.1 mL | |
Cellular Markers, Glia Markers, Neuroscience, Phospho Specific | |
3897 | |
Store at 4C in the dark. Do not freeze. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction