Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
L1CAM Antibody (UJ127.11), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1002682JF646
Description
L1CAM Monoclonal specifically detects L1CAM in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunofluorescence.Specifications
L1CAM | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
L1CAM | |
Homogenous suspension of 16 week human foetal brain. | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Flow Cytometry, Immunofluorescence, Immunohistochemistry | |
UJ127.11 | |
Flow Cytometry, Immunohistochemistry, Immunofluorescence | |
antigen identified by monoclonal R1, CAML1N-CAML1, CD171, CD171 antigen, HSAS, HSAS1, L1 cell adhesion molecule, MASA, MIC5, N-CAM-L1, NCAM-L1, neural cell adhesion molecule L1, S10, SPG1 | |
Mouse | |
IgG purified | |
Cellular Markers, Glia Markers, Neuroscience | |
3897 | |
Store at 4°C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction