Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
L1CAM Antibody (UJ127), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB7771B
Description
L1CAM Monoclonal antibody specifically detects L1CAM in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOFSpecifications
L1CAM | |
Monoclonal | |
Biotin | |
PBS | |
Mouse | |
Protein A or G purified from ascites | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunohistochemistry, CyTOF | |
UJ127 | |
Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
antigen identified by monoclonal R1, CAML1N-CAML1, CD171, CD171 antigen, HSAS, HSAS1, L1 cell adhesion molecule, MASA, MIC5, N-CAM-L1, NCAM-L1, neural cell adhesion molecule L1, S10, SPG1 | |
Homogenous suspension of 16 week human fetal brain., Accession # P32004 | |
0.1 mL | |
Cellular Markers, Glia Markers, Neuroscience | |
3897 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction