Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
L1CAM Antibody (UJ181.4), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1002683B
Description
L1CAM Monoclonal specifically detects L1CAM in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
L1CAM | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
L1CAM | |
16 week human foetal brain. | |
0.1 mL | |
Cellular Markers, Glia Markers, Neuroscience | |
3897 | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, Immunohistochemistry | |
UJ181.4 | |
Western Blot, Immunohistochemistry | |
antigen identified by monoclonal R1, CAML1N-CAML1, CD171, CD171 antigen, HSAS, HSAS1, L1 cell adhesion molecule, MASA, MIC5, N-CAM-L1, NCAM-L1, neural cell adhesion molecule L1, S10, SPG1 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction