Learn More
L1CAM/LI Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305304A

Description
The recommended shelf life for this product is 1 year from date of receipt.
The protein encoded by L1CAM is an axonal glycoprotein belonging to the immunoglobulin supergene family. The ectodomain, consisting of several immunoglobulin-like domains and fibronectin-like repeats (type III), is linked via a single transmembrane sequence to a conserved cytoplasmic domain. This cell adhesion molecule plays an important role in nervous system development, including neuronal migration and differentiation. Mutations in the gene cause X-linked neurological syndromes known as CRASH (corpus callosum hypoplasia, retardation, aphasia, spastic paraplegia and hydrocephalus). Alternative splicing of L1CAM results in multiple transcript variants, some of which include an alternate exon that is considered to be specific to neurons.Specifications
L1CAM/LI | |
Polyclonal | |
Unconjugated | |
L1CAM | |
NCAM-L1, SPG1, HSAS, N-CAM-L1, CAML1, MASA, N-CAML1, S10, CD171, L1 cell adhesion molecule, HSAS1, MIC5, L1CAM | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3897 | |
4° C | |
Liquid |
Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
P32004 | |
L1CAM | |
Between 1 and 50 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.