Learn More
L1TD1 Rabbit anti-Human, Polyclonal, Proteintech
SDP
Rabbit Polyclonal Antibody
Supplier:  Proteintech Group Inc 215281AP150UL
Encompass_Preferred
 View more versions of this product
Catalog No. 50-173-3467
Only null left
Add to Cart
Description
Description
L1TD1 is a member of the keratin gene family. As a type I hair keratin, it is an acidic protein which heterodimerizes with type II keratins to form hair and nails. The type I hair keratins are clustered in a region of chromosome 17q12-q21 and have the same direction of transcription.
Specifications
Specifications
L1TD1
Polyclonal
Unconjugated
L1TD1
ECAT11, ES cell associated protein 11, L1TD1
Rabbit
Antigen Affinity Chromatography
RUO
54596
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.38 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q5T7N2
L1TD1
L1TD1 Fusion Protein Ag16147
150 μL
Primary
Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.