Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
L2HGDH Mouse anti-Human, Porcine, Clone: 3D11C2, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
| Antigen | L2HGDH |
|---|---|
| Clone | 3D11C2 |
| Concentration | 1.32 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6702
![]()
|
Proteintech Group Inc
662691IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This gene encodes L-2-hydroxyglutarate dehydrogenase, a FAD-dependent enzyme that oxidizes L-2-hydroxyglutarate to alpha-ketoglutarate in a variety of mammalian tissues. Mutations in this gene cause L-2-hydroxyglutaric aciduria, a rare autosomal recessive neurometabolic disorder resulting in moderate to severe mental retardation.Specifications
| L2HGDH | |
| 1.32 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| C14orf160, DURANIN, L2HGDH | |
| L2HGDH | |
| IgG2a | |
| Protein A | |
| Antibody |
| 3D11C2 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Pig | |
| Q9H9P8 | |
| 100152180, 79944 | |
| L2HGDH Fusion Protein Ag8382 | |
| Primary | |
| -20°C | |
| L2HGDH |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title