Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ L2HGDH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564221
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564221 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564221 Supplier Invitrogen™ Supplier No. PA564221
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KKEPYCRGLM AIDCPHTGIV DYRQVALSFA QDFQEAGGSV LTNFEVKGIE MAKESPSRSI DGMQYPIVIK NTKGEEI Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 78%.

This gene encodes L-2-hydroxyglutarate dehydrogenase, a FAD-dependent enzyme that oxidizes L-2-hydroxyglutarate to alpha-ketoglutarate in a variety of mammalian tissues. Mutations in this gene cause L-2-hydroxyglutaric aciduria, a rare autosomal recessive neurometabolic disorder resulting in moderate to severe mental retardation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen L2HGDH
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene L2HGDH
Gene Accession No. Q9H9P8
Gene Alias 2-hydroxyglutarate dehydrogenase; alpha-hydroxyglutarate oxidoreductase; alpha-ketoglutarate reductase; BC016226; C14orf160; Duranin; FLJ12618; L2HGA; L2HGDH; L-2-hydroxyglutarate dehydrogenase; L-2-hydroxyglutarate dehydrogenase, mitochondrial; L-alpha-hydroxyglutarate dehydrogenase; RGD1306250
Gene Symbols L2HGDH
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human L2HGDH. Recombinant protein control fragment (Product #RP-104004).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79944
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.