Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L2HGDH Polyclonal Antibody, Invitrogen™

Catalog No. PIPA545638
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA545638 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA545638 Supplier Thermo Scientific Supplier No. PA545638

Rabbit Polyclonal Antibody

Peptide sequence: GSVLTNFEVK GIEMAKESPS RSIDGMQYPI VIKNTKGEEI RCQYVVTCAG Sequence homology: Cow: 91%; Dog: 86%; Guinea Pig: 86%; Horse: 86%; Human: 100%; Mouse: 79%; Pig: 100%; Rabbit: 86%

L2HGDH Polyclonal antibody specifically detects L2HGDH in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen L2HGDH
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene L2HGDH
Gene Accession No. Q9H9P8
Gene Alias 2-hydroxyglutarate dehydrogenase; alpha-hydroxyglutarate oxidoreductase; alpha-ketoglutarate reductase; BC016226; C14orf160; Duranin; FLJ12618; L2HGA; L2HGDH; L-2-hydroxyglutarate dehydrogenase; L-2-hydroxyglutarate dehydrogenase, mitochondrial; L-alpha-hydroxyglutarate dehydrogenase; RGD1306250
Gene Symbols L2HGDH
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human L2HGDH
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79944
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.