Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ L3HYPDH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113237
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113237 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113237 Supplier Invitrogen™ Supplier No. PA5113237
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

L3HYPDH Polyclonal Antibody for ELISA, IHC (P), Western Blot

L3HYPDH catalyzes the dehydration of trans-3-hydroxy-L-proline to delta-1-pyrroline-2-carboxylate (Pyr2C). It may be required to degrade trans-3-hydroxy-L-proline from the diet and originating from the degradation of proteins such as collagen-IV that contain it.
TRUSTED_SUSTAINABILITY

Specifications

Antigen L3HYPDH
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene L3HYPDH
Gene Accession No. Q96EM0
Gene Alias C14orf149; L-3-hydroxyproline dehydratase (trans-); L3HYPDH; probable proline racemase; trans-3-hydroxyl-L-proline dehydratase; trans-3-hydroxy-L-proline dehydratase; trans-L-3-hydroxyproline dehydratase
Gene Symbols L3HYPDH
Host Species Rabbit
Immunogen Recombinant Human Trans-3-hydroxy-L-proline dehydratase protein (13-312AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 112849
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.