Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ L3HYPDH Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563342
Description
Immunogen sequence: MESALAVPRL PPHDPGTPVL SVVDMHTGGE PLRIVLAGCP EVSGPTLLAK RRYMRQHLDH VRRRLMFEPR GHRDMYGAVL VPSEL Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 89%.
L3HYPDH catalyzes the dehydration of trans-3-hydroxy-L-proline to delta-1-pyrroline-2-carboxylate (Pyr2C). It may be required to degrade trans-3-hydroxy-L-proline from the diet and originating from the degradation of proteins such as collagen-IV that contain it.
Specifications
L3HYPDH | |
Polyclonal | |
Unconjugated | |
L3HYPDH | |
C14orf149; L-3-hydroxyproline dehydratase (trans-); L3HYPDH; probable proline racemase; trans-3-hydroxyl-L-proline dehydratase; trans-3-hydroxy-L-proline dehydratase; trans-L-3-hydroxyproline dehydratase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
112849 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96EM0 | |
L3HYPDH | |
Recombinant protein corresponding to Human L3HYPDH. Recombinant protein control fragment (Product #RP-101260). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction