Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
L3MBTL1 Antibody (RAB-C213), DyLight 488 - Chimeric, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP320137G
Description
L3MBTL1 Monoclonal antibody specifically detects L3MBTL1 in Human samples. It is validated for Flow Cytometry, ELISA, ImmunofluorescenceSpecifications
L3MBTL1 | |
Monoclonal | |
DyLight 488 | |
dJ138B7.3, FLJ41181, H-L(3)MBT, H-l(3)mbt protein, KIAA0681L3MBTLDKFZp586P1522, l(3)mbt (Drosophila)-like, L(3)mbt protein homolog, L(3)mbt-like, l(3)mbt-like (Drosophila), l(3)mbt-like 1 (Drosophila), L3MBT, lethal (3) malignant brain tumor l(3), lethal(3)malignant brain tumor-like protein | |
This antibody was obtained by recombinant antibody (rAb) phage display recognizing L3MBTL1 protein under non-denaturing conditions. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Flow Cytometry, ELISA, Immunofluorescence | |
RAB-C213 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
26013 | |
Store at 4°C in the dark. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction