Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
L3MBTL2 Antibody, DyLight 680, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP149965FR
Description
L3MBTL2 Polyclonal antibody specifically detects L3MBTL2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
L3MBTL2 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
dJ756G23.3, DKFZP761I141, H-l(3)mbt-l, H-l(3)mbt-like protein 2, l(3)mbt-like 2 (Drosophila), L(3)mbt-like protein 2, L3MBT, lethal(3)malignant brain tumor-like protein 2 | |
The immunogen for this product maps to a region between residue 655 and 705 of human Lethal (3) Malignant Brain Tumor-Like 2 using the numbering given in entry NP_113676.2 (GeneID 83746). | |
0.1 mL | |
Apoptosis, Cancer, Tumor Suppressors | |
83746 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 680 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction