Learn More
L3MBTL2 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302868A

Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predicted to react with Bovine and Orangutan
Putative Polycomb group (PcG) protein. PcG proteins maintain the transcriptionally repressive state of genes, probably via a modification of chromatin, rendering it heritably changed in its expressibility. Its association with a chromatin-remodeling complex suggests that it may contribute to prevent expression of genes that trigger the cell into mitosis. Binds to monomethylated and dimethylated ′Lys-20′ on histone H4. Binds histone H3 peptides that are monomethylated or dimethylated on ′Lys-4′, ′Lys-9′ or ′Lys-27′.Specifications
L3MBTL2 | |
Polyclonal | |
Unconjugated | |
L3MBTL2 | |
H-l(3)mbt-l, L3MBT, H-l(3)mbt-like protein 2, L(3)mbt-like protein 2, lethal(3)maligt brain tumor-like protein 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
83746 | |
4° C | |
Liquid |
Immunohistochemistry, Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q969R5 | |
L3MBTL2 | |
Between 655 and 705 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.