Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
L3MBTL3 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP147315AF750
Description
L3MBTL3 Polyclonal antibody specifically detects L3MBTL3 in Human samples. It is validated for Western BlotSpecifications
L3MBTL3 | |
Polyclonal | |
Western Blot | |
H-l(3)mbt-like protein 3, KIAA1798, l(3)mbt-like 3 (Drosophila), L(3)mbt-like protein 3, lethal(3)malignant brain tumor-like protein 3, MBT1, MBT-1, RP11-73O6.1 | |
The immunogen for this product maps to a region between residue 100 and 150 of human Lethal(3)Malignant Brain Tumor-like 3 using the numbering given in entry NP_115814.1 (GeneID 84456). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
84456 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction