Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ L3MBTL3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543622
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543622 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543622 Supplier Invitrogen™ Supplier No. PA543622
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MTESASSTSG QEFDVFSVMD WKDGVGTLPG SDLKFRVNEF GALEVITDEN Sequence homology: Cow: 100%; Dog: 93%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

Putative Polycomb group (PcG) protein. PcG proteins maintain the transcriptionally repressive state of genes, probably via a modification of chromatin, rendering it heritably changed in its expressibility.
TRUSTED_SUSTAINABILITY

Specifications

Antigen L3MBTL3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene L3MBTL3
Gene Accession No. Q96JM7
Gene Alias AI481284; H-l(3)mbt-like protein 3; KIAA1798; l(3)mbt-like 3; l(3)mbt-like 3 (Drosophila); l(3)mbt-like protein 3; L3MBTL histone methyl-lysine binding protein 3; L3mbtl3; L3MBTL3, histone methyl-lysine binding protein; lethal(3)malignant brain tumor-like protein 3; lethal(3)malignant brain tumor-like protein 3; LOW QUALITY PROTEIN: lethal(3)malignant brain tumor-like protein 3; MBT1; MBT-1
Gene Symbols L3MBTL3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human L3MBTL3 (aa 1-50).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84456
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.