Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
L3MBTL4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP215009
Description
L3MBTL4 Polyclonal specifically detects L3MBTL4 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
L3MBTL4 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
FLJ35936, H-l(3)mbt-like protein 4, HsT1031, l(3)mbt-like 4 (Drosophila), L(3)mbt-like protein 4, lethal(3)malignant brain tumor-like protein 4 | |
Rabbit | |
71 kDa | |
0.1 mL | |
Apoptosis, Cancer, Tumor Suppressors | |
91133 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot | |
1mg/mL | |
Western Blot 1:500-1:3000 | |
Q8NA19 | |
L3MBTL4 | |
Recombinant protein encompassing a sequence within the center region of human L3MBTL4. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction