Learn More
Siemens Healthineers LA Controls (US)
Click to view available options
Level:
High
Low
Description
LA Controls are positive controls for lupus anticoagulant clotting assays using LA 1 Screening and LA 2 Confirmation Reagents.
Specifications
Specifications
Type | Hemostasis Control |
For Use With (Application) | Thrombophilia Testing |
Level | Low |
Quantity | 6 x 1 mL |