Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ La Crosse Virus Monoclonal Antibody (10G5.4)
GREENER_CHOICE

Catalog No. 01671833
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-671-833 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-671-833 Supplier Invitrogen™ Supplier No. MA110804
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

MA1-10804 detects La Crosse Virus and recognizes G1 and G2. MA1-10804 has been successfully used in ELISA, Western blot, Immunofluorescence, immunoprecipitation, and neutralization applications. The MA1-10804 immunogen is La Crosse Virus grown on E6 Vero cells and purified by sucrose gradient centrifugation.

TRUSTED_SUSTAINABILITY

Specifications

Antigen La Crosse Virus
Applications ELISA, Immunoprecipitation, Neutralization, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 10G5.4
Concentration 1.3 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias LCV
Host Species Mouse
Immunogen La Crosse Virus grown on E6 Vero cells and purified by sucrose gradient centrifugation.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.