Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
La Crosse Virus Monoclonal Antibody (4A5.5), QED Bioscience™

Mouse Monoclonal Antibody
Supplier: Leinco Technologies Inc 18752100UG

This item is not returnable.
View return policy
Description
La Crosse Virus Monoclonal antibody specifically detects La Crosse Virus in Virus samples. It is validated for ELISA, Immunocytochemistry, Immunoprecipitation, Neutralization, Western BlotSpecifications
La Crosse Virus | |
Monoclonal | |
0.5-1.0 mg/mL | |
PBS with no preservative; pH 7.4 | |
Mouse | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG2a |
ELISA, Immunocytochemistry, Immunoprecipitation, Neutralization, Western Blot | |
4A5.5 | |
Unconjugated | |
LCV | |
La Crosse Virus (LACV) grown on E6 Vero cells and purified by sucrose gradient centrifugation. | |
100 μg | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction