Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
La Crosse Virus Nucleocapsid Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP241225MFV500
Description
La Crosse Virus Nucleocapsid Polyclonal antibody specifically detects La Crosse Virus Nucleocapsid in Virus samples. It is validated for ELISASpecifications
| La Crosse Virus Nucleocapsid | |
| Polyclonal | |
| ELISA | |
| LACVsSgp1, N, Nucleocapsid protein, Nucleoprotein, Protein N | |
| Antibody was raised against an 18 amino acid peptide near the center of the La Crosse virus nucleocapsid. The immunogen is located within amino acids 90 - 140 of La Crosse Virus Nucleocapsid. Amino Acid Squence: LDQYNENDDESQHELIRT | |
| 0.1 mL | |
| Infections (Virus Bacteria and Parasites) | |
| 956556 | |
| Store at 4C in the dark. | |
| IgG |
| ELISA | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction