Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lab Armor™ StayTemp™ Equipment Tray
SDP

Catalog No. p-4417887
Encompass
Change view
Click to view available options
Width (English):
6 in.
10 in.
Weight (English):
4 lb.
6 lb.
10 lb.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (English) Weight (English)
10-876-027 6 in. 6 lb.
10-876-028 6 in. 10 lb.
10-876-029 10 in. 4 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 10-876-027 Supplier Lab Armor™ Supplier No. 39956050
Only null left
Add to Cart
Add to Cart

For maintaining cold or heat with high efficiency in refrigerators, incubators, & ovens

StayTemp Equipment trays are an upgrade that keeps samples and reagents in your refrigerator, incubator, and laboratory ovens at temperature despite normal hourly fluctuations in air temperature.

  • Stainless steel construction
  • Keeps samples organized protecting sample from defrost cycles and constant door opening and closing
  • Eliminates the need of racks
  • Can be hand washed, autoclaved or towel dried
  • For any size and shape vessel including non-water-tight vessels
  • Available in 3 sizes

Does not include lid.

Specifications

Height (Metric) 10.16 cm
Length (Metric) 15.24 cm
Material Stainless Steel
Type Staytemp Equipment Tray
Width (Metric) 15.2 cm
For Use With (Application) For maintaining cold or heat with high efficiency in refrigerators, incubators and ovens
Includes 1 L lab armor beads
Length (English) 6 in.
Height (English) 4 in.
Temperature (English) 356°F
Weight (English) 6 lb.
Temperature (Metric) 180°C
Weight (Metric) 2.72 kg
Width (English) 6 in.
Autoclavable Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.