Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lab Performance Check, Method 200.7, Ricca Chemical

Click to view available options
Quantity:
100 mL
500 mL
Specifications
Specifications
Packaging | Amber Poly Bottle |
CAS Min % | 91.0, 6.5, 0.05, 0.03, 0.02, 0.01 |
CAS Max % | 94.0, 6.76, 0.05, 0.03, 0.02, 0.02 |
CAS | 7697-37-2, 16919-19-0, 7722-76-1, 7784-27-2, 584-08-7 |
Methods | Method 200.7 |
Physical Form | Liquid |
Melting Point | Approximately 0°C |
Quantity | 100 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction