Learn More
Thermo Scientific™ Lab Rotators
Rotate flasks, test tube racks, beakers, vials, Petri dishes, microwell plates, culture plates, plastic/glass trays and slides with these lab rotors.
Supplier:  Thermo Scientific™ 2314Q
 View more versions of this product
Catalog No. 11-671-52Q
Description
Description
Rotate flasks, test tube racks, beakers, vials, Petri dishes, microwell plates, culture plates, plastic/glass trays and slides in microbiological, immunological and general clinical applications with Thermo Scientific™ Lab Rotators. Improved drive eccentrics and an upgraded motor ensures stability, enhanced long-term performance and quiet, dependable operation. For use in incubators, warm rooms, environmental chambers and refrigerators.
Features:
  • Two platform sizes available
  • Continuous-duty motor provides smooth, quiet orbit
  • Timer makes it easy to perform time studies; set for continuous or timed
  • Autoclavable white silicone nonslip platform mat cleans easily
  • Variable speed control for gentle or vigorous agitation
Warranty:
  • One year parts, 90 days labor
TRUSTED_SUSTAINABILITY
Specifications
Specifications
19 mm
40 to 220 rpm
120 V
50 W
Rotator
UL
50/60 Hz
4.38 in.
13.62 in.
35 cm
12.5 in.
12 in.
16 lb.
20 to 80%
4.5 kg
120 V, 50/60 Hz
4°C to 40°C
0.4 A
10 lb.
39.2° to 104°F
13.62 x 12.5 x 4.38 in. (35 x 32 x 11 cm)
11 cm
12 in.
30 cm
12.5 in.
30 cm
7.3 kg
1 to 60 min.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.