Learn More
United Scientific™ Lab Scoop And Handle Set
SDPSCI_ED
Catalog No. S15686 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Includes:
Handle Only
Scoop Only
Scoop and Handle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
S15686 Scoop Only
S15809 Scoop and Handle
S15728 Handle Only
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S15686 Supplier United Scientific™ Supplier No. LSCSS6
Only null left
Add to Cart
Add to Cart
Ideal for scooping powders in the lab
  • Stainless steel scoop with one pointed end for handling small amounts of substances.
  • Includes sturdy hardwood handle
  • Length of scoop is 6.5 in. (16.5 cm)
  • Handle measures 5 in. (12.7 cm) long
  • Available as a set or handles and scoops separately.
Specifications
Sterility Non-sterile
Autoclavable Autoclavable
Product Type Scoop
Includes Scoop Only
Material Stainless Steel