Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ 14.3.3σ Ab-1 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Catalog No. MS1185PABX Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
Prediluted
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS1185PABX 200 μL Concentrated 1mg/mL
MS1185P1ABX 100 μL Concentrated 1mg/mL
MS1185P1 500 μL Concentrated 200μg/mL
MS1185P 1 mL Concentrated 200μg/mL
MS1185R7 7 mL Prediluted Prediluted
MS1185P0 100 μL Concentrated 200μg/mL
MS1185R1 1 mL Prediluted Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. MS1185PABX Supplier Epredia™ Supplier No. MS1185PABX

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Take advantage of a quality product that delivers accurate results with the Epredia™ 14.3.3σ Ab-1, Mouse Monoclonal Antibody.

14.3.3 proteins are a family of small, widely expressed, highly conserved cytosolic proteins. 14.3.3 proteins bind to and influence the activities of a diverse group of molecules involved in signal transduction, cell cycle regulation and apoptosis, including Raf, PKC, Bad, Cbl, and c-Bcr. Interactions between 14.3.3 and target proteins are strongly influenced by the phosphorylation state of 14.3.3 and the target protein. 14.3.3 σ shares cyclin-Cdk2 binding motifs with different cell cycle regulators, including p107, p130, p21Cip1, p27Kip1, and p57Kip2 and is associated with cyclin-Cdk complexes in vitro and in vivo.

Host Species: Mouse

Clone: 1433S01

Isotype: IgG1/κ

Species Reactivity: Human. Others not tested.

Epitope: Not determined

Immunogen: Recombinant human 14.3.3σ protein

Molecular Weight: 26-30kDa

Positive Control: HT29 cells, Skin

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen 14.3.3sigma Ab-1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 1433S01
Concentration 1mg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant human 1433 sigma protein
Quantity 200 μL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.