Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Actin, Skeletal Muscle, Ab-2 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS185P Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS185P 1 mL Concentrated
MS185R7 7 mL Prediluted
MS185P0 100 μL Concentrated
MS185P1 500 μL Concentrated
MS185R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS185P Supplier Epredia Supplier No. MS185P

Obtain accurate, reproducible results in immunohistochemistry and immunofluorescence experiments with Epredia™ Actin Skeletal Muscle Ab-2 Mouse Monoclonal Antibody.

Host Species: Mouse

Clone: 5C5.F8.C7 (α-Sr-1)

Isotype: IgM/κ

Species Reactivity: Human, Cow, Rabbit, Sheep, Rat, Guinea Pig and Frog. Others not known.

Epitope: N-terminal decapeptide

Immunogen: N-Terminal decapeptide of α skeletal muscle isoform of actin; acetylated at the N-terminus

Positive Control: Skeletal or Cardiac muscle, or Rhabdomyosarcoma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Actin, Skeletal Muscle Ab-2
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 5C5.F8.C7; or alpha-Sr-1
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen N-terminal decapeptide
Quantity 1 mL
Regulatory Status RUO
Research Discipline Muscle Biology
Primary or Secondary Primary
Target Species Bovine, Frog, Guinea Pig, Human, Rabbit, Rat, Sheep
Form Concentrated
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.