Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Actin, Smooth Muscle, Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS113B Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
12 mL
Conjugate:
Biotin
Unconjugated
Form:
Concentrated
Prediluted
Ready-to-Use
Concentration:
1mg/mL
40μg/mL
Prediluted
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Form Concentration
MS113B 1 mL Biotin Concentrated 40μg/mL
MS113B1 500 μL Biotin Concentrated 40μg/mL
MS113P1 500 μL Unconjugated Concentrated 40μg/mL
MS113P 1 mL Unconjugated Concentrated 40μg/mL
MS113R7 7 mL Unconjugated Prediluted Prediluted
MS113RQ 12 mL Unconjugated Ready-to-Use Prediluted
MS113B0 100 μL Biotin Concentrated 40μg/mL
MS113P1ABX 100 μL Unconjugated Concentrated 1mg/mL
MS113PABX 200 μL Unconjugated Concentrated 1mg/mL
MS113R1 1 mL Unconjugated Prediluted Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. MS113B Supplier Epredia Supplier No. MS113B

Obtain accurate, reproducible results in immunohistochemistry and immunofluorescence experiments with Epredia™ Actin, Smooth Muscle Ab-1, Mouse Monoclonal Antibody.

Host Species: Mouse

Clone: 1A4; also known as asm-1

Isotype: IgG2a/κ

Species Reactivity: Human, Baboon, Cow, Rabbit, Mouse, Rat and Chicken. Others not known.

Epitope: Acetyl group and the first four amino acids on the N-terminal end of the peptidic chain of α-smooth actin

Immunogen: N-terminal decapeptide of α-smooth muscle isoform of actin; acetylated at the N-terminus

Positive Control: Smooth muscle or leiomyosarcoma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence

Specifications

Antigen Actin, Smooth Muscle Ab-1
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1A4 ; also known as asm-1
Concentration 40μg/mL
Conjugate Biotin
Host Species Mouse
Immunogen N-terminal decapeptide of alpha-smooth muscle isoform of actin; acetylated at the N-terminus
Quantity 1 mL
Regulatory Status IVD
Research Discipline Muscle Biology
Primary or Secondary Primary
Target Species Bovine, Chicken, Human, Mouse, Non-human Primate, Rabbit, Rat
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.