Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Actin, Smooth Muscle, Ab-1 Mouse Monoclonal Antibody, 40μg/mL, BSA and azide

Catalog No. MS1143P1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Concentration:
1mg/mL
200μg/mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
MS1143P1 500 μL 200μg/mL
MS1143P0 100 μL 200μg/mL
MS1143P 1 mL 200μg/mL
MS1143P1ABX 100 μL 1mg/mL
MS1143PABX 200 μL 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS1143P1 Supplier Epredia™ Supplier No. MS1143P1

Ensure accurate, reproducible results in western blotting and immunoprecipitation experiments with Epredia™ Caspase 8 (FLICE) Ab-3, Mouse Monoclonal Antibody.

Caspase 8 binds to the death effector domain (DED) of FADD through an analogous DED domain present in tandem in the proform of the caspase 8 protein. Recruitment of caspase 8 to the Fas receptor results in oligomerization of the caspase 8 protein, which in turn drives its autoactivation through “self-cleavage”. Activated caspase 8 then activates other downstream caspases including caspase 9, thereby commiting the cell to undergo apoptosis.

Host Species: Mouse

Clone: 8CSP03; same as 4.1.20

Isotype: IgG1

Species Reactivity: Human. Others not tested.

Epitope: Not determined

Immunogen: Recombinant full-length human Caspase 8 protein

Molecular Weight: 55 kDa

Positive Control: Jurkat cells

Cellular Localization: Cytoplasmic

Recommended for:

  • Western Blotting
  • Immunoprecipitation (Denatured verified)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Caspase 8 (FLICE) Ab-3
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 8CSP03; same as 4.1.20
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant full length human Caspase 8 protein
Quantity 500 μL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.