Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Adenovirus Type 2 and 5 Ab-5 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Catalog No. MS1069R7 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS1069R7 7 mL Prediluted -
MS1069P1ABX 100 μL Concentrated 1mg/mL
MS1069PABX 200 μL Concentrated 1mg/mL
MS1069P0 100 μL Concentrated 200μg/mL
MS1069P 1 mL Concentrated 200μg/mL
MS1069R1 1 mL Prediluted -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. MS1069R7 Supplier Epredia™ Supplier No. MS1069R7

Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, and immunoprecipitation experiments.

Adenovirus Ab-5 detects adenovirus serogroups 2 and 5. The adenovirus early gene product E1A is a potent stimulator of cellular proliferation, which when overexpressed can overcome the growth-inhibitory effects of TGF-β. The E1A region encodes a series of related proteins (35-46kDa) with multifuctional capabilities and form a specific complex with the retinoblastoma tumor suppressor gene product (Rb protein). The E1A and E1B regions together comprise the transforming region of adenovirus. While expression of E1A alone is sufficient to immortalize primary cells, complete transformation requires the additional expression of the E1B region. Several conserved regions of E1A are similar to portions of other viral oncoproteins like the HPV-16 and HPV-18 E7 and SV40 large T antigen.

Host Species: Mouse

Clone: M58 + M73

Isotype: IgG2a/κ + IgG2a/κ

Species Reactivity: Adenovirus.

Immunogen: Adenovirus

Positive Control: 293 cells or adenovirus infected tissues

Cellular Localization: Nuclear.

Ab-5 is excellent for staining of formalin-fixed, paraffin-embedded tissues.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native verified)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Adenovirus Type 2 and 5 Ab-5
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone M58 + M73
Conjugate Unconjugated
Host Species Mouse
Immunogen Adenovirus
Quantity 7 mL
Regulatory Status RUO
Research Discipline Infectious Disease
Primary or Secondary Primary
Target Species Adenovirus
Form Prediluted
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.