Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ ALK (Anaplastic Lymphoma Kinase)/p80 Rabbit Monoclonal Antibody

Catalog No. RM9108RQ Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
1 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
RM9108RQ 12 mL Ready-to-Use
RM9108S0 100 μL Concentrated
RM9108R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. RM9108RQ Supplier Epredia™ Supplier No. RM9108RQ

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ ALK (Anaplastic Lymphoma Kinase)/p80, Rabbit Monoclonal Antibody.

Recognizes a human p80 protein, identified as a hybrid of the anaplastic lymphoma kinase (ALK) gene and the nucleophosmin (NPM) gene resulting from the t(2;5)(p23;q35) translocation found in 30-50% of CD30+ large cell lymphomas (1). This rabbit monoclonal antibody can be used to detect p80 in these lymphomas and may also be used to detect a recently described subtype of large B cell lymphoma, which expresses the full-length ALK protein (2).

Host Species: Rabbit

Clone: SP8

Isotype: IgG

Species Reactivity: Human. Others not tested.

Epitope: Not determined

Immunogen: Recombinant protein corresponding to a region which spans the tyrosine kinase catalytic domain and part of the C-terminus of the NPM-ALK transcript

Positive Control: Anaplastic lymphoma

Cellular Localization: Cytoplasmic + nuclear Clone SP8 is excellent for staining of formalin/paraffin with no special pretreatment

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen ALK (Anaplastic Lymphoma Kinase)/p80
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SP8
Conjugate Unconjugated
Host Species Rabbit
Immunogen Recombinant protein corresponding to a region, which spans the tyrosine kinase catalytic domain and part of the C-terminus of the NPM-ALK transcript
Quantity 12 mL
Regulatory Status IVD
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Form Ready-to-Use
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.