Learn More
Description
Host Species: Mouse
Clone: mcl
Isotype: IgG2a/κ
Species Reactivity: Human. Others not known.
Immunogen: Human amyloid A
Positive Control: Amyloid deposits in kidney
Cellular Localization: Extracellular
Ab-1 reacts with isolated protein AA in both native fibril form and denatured form. It reacts with amyloid deposits in all organs and tissues including kidney and rectum, and as well as the serum precursor of protein AA.
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Amyloid A Component Ab-1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | mcl |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Human amyloid A |
| Quantity | 7 mL |
| Regulatory Status | RUO |
| Research Discipline | Neuroscience |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.