Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Amyloid A Component Ab-1 Mouse Monoclonal Antibody

Catalog No. MS1219S0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS1219S0 100 μL Concentrated
MS1219S1 500 μL Concentrated
MS1219S 1 mL Concentrated
MS1219R1 1 mL Prediluted
MS1219R7 7 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS1219S0 Supplier Epredia™ Supplier No. MS1219S0

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Amyloid A Component Ab-1, Mouse Monoclonal Antibody.

Host Species: Mouse

Clone: mcl

Isotype: IgG2a/κ

Species Reactivity: Human. Others not known.

Immunogen: Human amyloid A

Positive Control: Amyloid deposits in kidney

Cellular Localization: Extracellular

Ab-1 reacts with isolated protein AA in both native fibril form and denatured form. It reacts with amyloid deposits in all organs and tissues including kidney and rectum, and as well as the serum precursor of protein AA.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Amyloid A Component Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone mcl
Conjugate Unconjugated
Host Species Mouse
Immunogen Human amyloid A
Quantity 100 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.