Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amyloid Precursor Protein (APP) Rabbit Polyclonal Antibody, Epredia™

Catalog No. RB9023P Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 mL
Form:
Concentrated
Prediluted
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
RB9023P 1 mL Concentrated
RB9023R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RB9023P Supplier Epredia Supplier No. RB9023P

Obtain accurate, reproducible results in immunohistochemistry and western blotting experiments.

Recognizes APP (b-APP770), MW 110kDa. Predicted to recognize Β-APP695 and Β-APP751. Amyloid precursor protein (APP) and APP-like proteins are transmembrane glycoproteins with a similar modular domain structure.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse and Rat. Others not tested.

Epitope: N-terminal

Immunogen: A synthetic peptide corresponding to N-terminal of human APP

Molecular Weight: 110kDa

Positive Control: Brain

Cellular Localization: Cytoplasmic and extracellular

This antibody is excellent for staining of formalin-fixed, paraffin-embedded tissues

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Specifications

Antigen Amyloid Precursor Protein (APP)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
Immunogen A synthetic peptide corresponding to N-terminal of human APP
Quantity 1 mL
Regulatory Status IVD
Research Discipline Neuroscience
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Form Concentrated
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.