Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Androgen Receptor Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS443P0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
12 mL
Conjugate:
Biotin
Unconjugated
Form:
Concentrated
Prediluted
Ready-to-Use
Concentration:
1mg/mL
200μg/mL
Prediluted
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Form Concentration
MS443P0 100 μL Unconjugated Concentrated 200μg/mL
MS443P1 500 μL Unconjugated Concentrated 200μg/mL
MS443B0 100 μL Biotin Concentrated 200μg/mL
MS443B 1 mL Biotin Concentrated 200μg/mL
MS443P1ABX 100 μL Unconjugated Concentrated 1mg/mL
MS443PABX 200 μL Unconjugated Concentrated 1mg/mL
MS443P 1 mL Unconjugated Concentrated 200μg/mL
MS443R1 1 mL Unconjugated Prediluted Prediluted
MS443R7 7 mL Unconjugated Prediluted Prediluted
MS443RQ 12 mL Unconjugated Ready-to-Use Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. MS443P0 Supplier Epredia Supplier No. MS443P0

Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, immunoprecipitation, and gel supershift experiments.

The expression of AR is reportedly inversely correlated with histologic grade, i.e. well differentiated prostate tumors show higher expression than the poorly differentiated tumors. In prostate cancer, AR has been proposed as a marker of hormone-responsiveness.

Host Species: Mouse

Clone: AR 441

Isotype: IgG1

Species Reactivity: Human and Dog. Does not react with mouse. Others not tested.

Epitope: aa 299-315

Immunogen: A synthetic peptide from human AR

Molecular Weight: 110kDa

Positive Control: LnCAp cells or prostate CA

Cellular Localization: Nuclear

Ab-1 is excellent for staining of formalin/paraffin tissues.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Denatured verified)
  • Gel Supershift

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Androgen Receptor Ab-1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone AR 441
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen A synthetic peptide from human AR
Quantity 100 μL
Regulatory Status RUO
Research Discipline Steroids Hormones and Growth Factors
Primary or Secondary Primary
Target Species Canine, Human
Form Concentrated
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.