Learn More
Description
Host Species: Mouse
Clone: AR 441
Isotype: IgG1
Species Reactivity: Human and Dog. Does not react with mouse. Others not tested.
Epitope: aa 299-315
Immunogen: A synthetic peptide from human AR
Molecular Weight: 110kDa
Positive Control: LnCAp cells or prostate CA
Cellular Localization: Nuclear
Ab-1 is excellent for staining of formalin/paraffin tissues.
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Denatured verified)
- Gel Supershift
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Androgen Receptor Ab-1 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | AR 441 |
| Concentration | Prediluted |
| Conjugate | Unconjugated |
| Formulation | Ready-to-use for IHC (UVLP) |
| Host Species | Mouse |
| Immunogen | A synthetic peptide from human AR |
| Quantity | 1 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.