Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Androgen Receptor (pSer216) Rabbit Polyclonal Antibody with BSA and azide

Stain formalin-fixed, paraffin-embedded tissues with Epredia™ Androgen Receptor, Rabbit Polyclonal Antibody.

Supplier:  Epredia™ RB9030P

 View more versions of this product

Catalog No. RB9030P


Description

Description

The androgen receptor (AR) is a member of the steroid superfamily of ligand-dependent transcription factors. The expression of AR is reportedly inversely correlated with histologic grade, i.e. well-differentiated prostate tumors show higher expression than the poorly differentiated tumors. In prostate cancer, AR has been proposed as a marker of hormone-responsiveness.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse and Rat. Others not tested.

Epitope: N-terminal

Immunogen: A synthetic peptide derived from N-terminal of rat AR

Molecular Weight: 110kDa

Positive Control: LnCAp cells or prostate CA

Cellular Localization: Nuclear

This antibody is suitable for staining of formalin-fixed, paraffin-embedded tissues.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation
Specifications

Specifications

Androgen Receptor
Polyclonal
200µg/mL
Rabbit
1 mL
Oncogenesis
Human, Mouse
IgG
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Polyclonal
Unconjugated
A synthetic peptide derived from N-terminal of rat AR
IVD
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.