Learn More
Description
Host Species: Mouse
Clone: 2D2
Isotype: IgG1/κ
Species Reactivity: Human. Does not react with mouse and rat. Others not tested.
Epitope: aa 3-16
Immunogen: A synthetic peptide, aa 3-16 (Cys-GSGEQPRGGGPTSS) of human Bax protein. This amino acid sequence is not shared by mouse and rat Bax protein.
Molecular Weight: 21kDa
Positive Control: HeLa cells, Hodgkin's lymphoma
Cellular Localization: Cytoplasmic and cell membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Flow Cytometry (Not tested)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Bax Ab-1 |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | 2D2 |
| Concentration | 200μg/mL |
| Conjugate | Biotin |
| Host Species | Mouse |
| Immunogen | A synthetic peptide, aa 3-16 (Cys-GSGEQPRGGGPTSS) of human bax protein. This amino acid sequence is NOT shared by mouse and rat bax protein. |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.